Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hrasls5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025731.2 | NP_080007.1 | 3 | 409 | ||
GenBank mRNA | AB261168.1 | - | 3 | 367 | ||
AB447517.1 | - | 3 | 367 | |||
AK006527.1 | - | 3 | 374 | |||
AK006790.1 | - | 3 | 374 | |||
AK007015.1 | - | 3 | 440 | |||
AK014971.1 | - | 3 | 410 | |||
BC061008.1 | - | 3 | 389 | 6772458 |