Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ormdl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025661.4 | NP_079937.1 | 4 | 659 | ||
GenBank mRNA | AK009621.1 | - | 4 | 660 | ||
AK012718.1 | - | 4 | 629 | |||
AK087497.1 | - | 4 | 628 | |||
AK154085.1 | - | 4 | 651 | |||
BC026412.1 | - | 2 | 334 | |||
BC046594.1 | - | 4 | 597 | 6308578 |