Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lonp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025827.3 | NP_080103.1 | 2 | 484 | ||
GenBank mRNA | AK004843.1 | - | 2 | 398 | ||
AK050231.1 | - | 2 | 393 | |||
AK088348.1 | - | 2 | 485 | |||
AK151103.1 | - | 2 | 430 | |||
AK151406.1 | - | 2 | 424 | |||
AK152964.1 | - | 2 | 424 | |||
AK168628.1 | - | 2 | 434 | |||
AK168791.1 | - | 2 | 430 | |||
AK168956.1 | - | 2 | 430 | |||
AK169656.1 | - | 398 | ||||
BC049090.1 | - | 2 | 420 | 6401542 |