Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027349.3 | NP_081625.3 | 14 | 1799 | ||
GenBank mRNA | AK011184.1 | - | 2 | 154 | ||
AK087936.1 | - | 2 | 124 | |||
AK142132.1 | - | 8 | 1075 | |||
AK162135.1 | - | 1897 | ||||
BC066150.1 | - | 3 | 233 | |||
BC067400.1 | - | 3 | 233 | |||
BC158104.1 | - | 13 | 1679 | 9007388 |