Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Higd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025933.3 | NP_080209.1 | 1 | 142 | ||
GenBank mRNA | AK005269.1 | - | 1 | 143 | ||
AK008387.1 | - | 1 | 139 | |||
BC021471.1 | - | 1 | 139 | 5343356 |