Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000159.3 | NP_000150.1 | 10 | 1099 | ||
NM_013976.3 | NP_039663.1 | 10 | 1099 | |||
NR_102316.1 | 10 | 1154 | ||||
NR_102317.1 | 9 | 1372 | ||||
XM_006722721.2 | XP_006722784.1 | 10 | 1068 | |||
XM_011527899.2 | XP_011526201.1 | 10 | 1068 | |||
XM_011527900.2 | XP_011526202.1 | 10 | 1068 | |||
XM_017026580.1 | XP_016882069.1 | 10 | 1068 | |||
GenBank mRNA | AK098370.1 | - | 10 | 1068 | ||
AK290407.1 | - | 10 | 1068 | |||
AK296446.1 | - | 10 | 1117 | |||
AK298772.1 | - | 8 | 969 | |||
AK300841.1 | - | 9 | 1321 | |||
BC002579.2 | - | 10 | 1052 | 3142978 | ||
BT006706.1 | - | 9 | 991 | |||
U69141.1 | - | 9 | 1027 |