Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025900.2 | NP_080176.2 | 2, 3 | 313 | ||
GenBank mRNA | AK012615.1 | - | 2, 3 | 254 | ||
AK012877.1 | - | 2, 3 | 248 | |||
AK033451.1 | - | 2, 3 | 203 | |||
AK154013.1 | - | 2, 3 | 295 | |||
BC048844.1 | - | 2, 3 | 263 | |||
BC055451.1 | - | 2, 3 | 259 | 4164972 | ||
BU920290.1 | - | 2, 3 | 297 |