Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027349.3 | NP_081625.3 | 14 | 1852 | ||
GenBank mRNA | AK011184.1 | - | 2 | 207 | ||
AK087936.1 | - | 2 | 177 | |||
AK142132.1 | - | 8 | 1128 | |||
AK162135.1 | - | 1950 | ||||
BC066150.1 | - | 3 | 286 | |||
BC067400.1 | - | 3 | 286 | |||
BC158104.1 | - | 13 | 1732 | 9007388 |