Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025895.4 | NP_080171.1 | 4 | 762 | ||
GenBank mRNA | AK003854.1 | - | 4 | 675 | ||
AK005130.1 | - | 1 | 170 | |||
AK005681.1 | - | 4 | 671 | |||
AK007855.1 | - | 4 | 691 | |||
AK154249.1 | - | 4 | 688 | |||
BC024941.1 | - | 4 | 705 | 4017146 |