Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam103a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025997.2 | NP_080273.1 | 2 | 262 | ||
GenBank mRNA | AK010694.1 | - | 2 | 218 | ||
AK011881.1 | - | 2 | 262 | |||
AK018614.1 | - | 2 | 261 | |||
AK079819.1 | - | 2 | 241 | |||
AK081301.1 | - | 2 | 254 | |||
AK085230.1 | - | 2 | 260 | |||
AK150938.1 | - | 2 | 229 | |||
AK160347.1 | - | 2 | 261 | |||
AK169392.1 | - | 2 | 291 | |||
BC058367.1 | - | 2 | 185 | 6830048 | ||
BC091760.1 | - | 2 | 222 | 5042613 | ||
BC096399.1 | - | 2 | 220 | 876882 |
Gene Symbol: Gm10767 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177750.1 | NP_001171221.1 | 3 | 561 | ||
GenBank mRNA | AK139034.1 | - | 3 | 562 | ||
AK163421.1 | - | 908 |