Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam69a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026062.4 | NP_080338.1 | 2 | 249 | ||
GenBank mRNA | AK008504.1 | - | 2 | 192 | ||
AK013580.1 | - | 2 | 213 | |||
AK038811.1 | - | 2 | 206 | |||
AK048979.1 | - | 2 | 206 | |||
AK050305.1 | - | 2 | 206 | |||
AK053273.1 | - | 1 | 54 | |||
AK153488.1 | - | 2 | 204 | |||
BC055842.1 | - | 2 | 181 | |||
BC061013.1 | - | 2 | 201 | |||
BC095935.1 | - | 2 | 183 | |||
BC116683.1 | - | 2 | 194 | 40106565 | ||
BY131147.1 | - | 2 | 251 |