Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyzl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026092.3 | NP_080368.1 | 3 | 347 | ||
GenBank mRNA | AK006241.1 | - | 3 | 348 | ||
AK006630.1 | - | 3 | 348 | |||
AY572447.1 | - | 3 | 306 | |||
BC048450.1 | - | 3 | 318 | 6774504 |