Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Herc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026101.4 | NP_080377.2 | 3 | 561 | ||
NM_030114.2 | NP_084390.1 | 3 | 561 | |||
GenBank mRNA | AK009436.1 | - | 3 | 416 | ||
AK019546.1 | - | 3 | 555 | |||
AK143111.1 | - | 3 | 435 | |||
AK147723.1 | - | 3 | 448 | |||
BC043082.1 | - | 3 | 373 | 6417111 | ||
BC060033.1 | - | 1 | 146 | 6478123 |