Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kbtbd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025991.3 | NP_080267.1 | 2 | 281 | ||
GenBank mRNA | AK010998.1 | - | 2 | 282 | ||
AK014373.1 | - | 2 | 207 | |||
BC025103.1 | - | 2 | 186 | 4481431 |