Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam103a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025997.2 | NP_080273.1 | 3 | 335 | ||
GenBank mRNA | AK010694.1 | - | 3 | 291 | ||
AK011881.1 | - | 3 | 335 | |||
AK018614.1 | - | 3 | 334 | |||
AK079819.1 | - | 3 | 314 | |||
AK081301.1 | - | 3 | 327 | |||
AK085230.1 | - | 3 | 333 | |||
AK150938.1 | - | 3 | 302 | |||
AK160347.1 | - | 3 | 334 | |||
AK169392.1 | - | 3 | 364 | |||
BC058367.1 | - | 3 | 258 | 6830048 | ||
BC091760.1 | - | 3 | 295 | 5042613 | ||
BC096399.1 | - | 3 | 293 | 876882 |