Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gemin6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026053.3 | NP_080329.1 | 2 | 300 | ||
GenBank mRNA | AK011454.1 | - | 2 | 281 | ||
AK013396.1 | - | 2 | 302 | |||
AK018385.1 | - | 2 | 297 | |||
AK020147.1 | - | 2 | 324 | |||
AK157745.1 | - | 2 | 313 | |||
AK167680.1 | - | 2 | 293 | |||
AK167920.1 | - | 2 | 339 | |||
BC025157.1 | - | 2 | 283 | 3670793 |