Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011416.2 | NP_035546.2 | 32 | 4705 | ||
NM_026003.2 | NP_080279.1 | 8 | 854 | |||
GenBank mRNA | AK009309.1 | - | 4 | 364 | ||
AK011935.1 | - | 8 | 799 | |||
AK147322.1 | - | 32 | 4854 | |||
BC004653.1 | - | 4 | 352 | |||
BC022975.1 | - | 10 | 1331 | |||
BC056199.1 | - | 22 | 2803 | |||
BC075641.1 | - | 32 | 4687 | 30544092 |