Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026009.2 | NP_080285.2 | 11, 12 | 1448 | ||
GenBank mRNA | AB040489.1 | - | 11, 12 | 1425 | ||
AK011882.1 | - | 11, 12 | 1385 | |||
AK078812.1 | - | 11, 12 | 1427 | |||
AK132273.1 | - | 11, 12 | 1406 | |||
AK162160.1 | - | 4, 5 | 308 | |||
BC120502.1 | - | 11, 12 | 1336 | 40129425 | ||
BC137681.1 | - | 11, 12 | 1336 | 8860700 |