Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026034.4 | NP_080310.1 | 6 | 932 | ||
GenBank mRNA | AK009821.1 | - | 6 | 926 | ||
AK011303.1 | - | 6 | 931 | |||
AK012872.1 | - | 6 | 907 | |||
AK015272.1 | - | 6 | 1065 | |||
BC038487.1 | - | 6 | 884 | 2812558 | ||
BC046281.1 | - | 6 | 884 | 6395384 | ||
BC058573.1 | - | 6 | 973 | 6487440 |