Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dock7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290636.1 | NP_001277565.1 | 49 | 6605 | ||
NM_026082.5 | NP_080358.3 | 48 | 6515 | |||
GenBank mRNA | AK014226.1 | - | 5 | 718 | ||
AK122554.2 | - | 18 | 2249 | |||
BC024823.1 | - | 19 | 2617 | |||
BC024917.1 | - | 13 | 1770 | |||
BC055032.1 | - | 8 | 1011 | |||
BC086672.1 | - | 29 | 4023 | 30647593 | ||
BC094420.1 | - | 37 | 4806 |