Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paxbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026110.2 | NP_080386.3 | 10 | 1702 | ||
GenBank mRNA | AK007365.1 | - | 4 | 407 | ||
AK079394.1 | - | 3 | 566 | |||
AY033907.1 | - | 10 | 1487 | |||
AY033908.1 | - | 10 | 1504 | |||
BC014838.1 | - | 3 | 187 | 3588322 | ||
BC027145.1 | - | 2 | 122 |