Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dedd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207677.3 | NP_997560.3 | 5 | 1293 | ||
GenBank mRNA | AK010701.1 | - | 5 | 1105 | ||
AK011713.1 | - | 4 | 999 | |||
AK043908.1 | - | 1138 | ||||
AK089096.1 | - | 3 | 664 | |||
BC037043.1 | - | 4 | 919 | |||
BC092250.1 | - | 5 | 1293 | 5043318 |