Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isoc2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026158.2 | NP_080434.1 | 2 | 202 | ||
GenBank mRNA | AK002905.1 | - | 2 | 203 | ||
AK007698.1 | - | 2 | 159 | |||
BC046756.1 | - | 2 | 174 | 5693399 | ||
BC049278.1 | - | 2 | 189 | 4485022 |