Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026222.2 | NP_080498.1 | 1 | 190 | ||
GenBank mRNA | AK014839.1 | - | 1 | 182 | ||
AK029948.1 | - | 1 | 146 | |||
AK030087.1 | - | 1 | 157 | |||
AK162849.1 | - | 1 | 191 | |||
AK163953.1 | - | 1 | 160 | |||
BC138917.1 | - | 1 | 152 | 8861939 | ||
BC138920.1 | - | 1 | 152 | 8861942 |