Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qpctl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026111.3 | NP_080387.2 | 4, 5 | 802 | ||
GenBank mRNA | AK007555.1 | - | 4, 5 | 847 | ||
AK007701.1 | - | 4, 5 | 830 | |||
AK035587.1 | - | 4, 5 | 802 | |||
AK045687.1 | - | 4, 5 | 806 | |||
BC058181.1 | - | 4, 5 | 825 | 5325516 |