Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qpctl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026111.3 | NP_080387.2 | 7 | 1193 | ||
GenBank mRNA | AK007555.1 | - | 7 | 1238 | ||
AK007701.1 | - | 7 | 1221 | |||
AK035587.1 | - | 7 | 1193 | |||
AK045687.1 | - | 7 | 1197 | |||
AK136746.1 | - | 335 | ||||
AK205714.1 | - | 1 | 66 | |||
BC058181.1 | - | 7 | 1216 | 5325516 |