Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agpat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026212.2 | NP_080488.1 | 5, 6 | 876 | ||
GenBank mRNA | AK010891.1 | - | 5, 6 | 686 | ||
AY072769.1 | - | 5, 6 | 669 | |||
BC125530.1 | - | 5, 6 | 750 | 40130045 | ||
BC137840.1 | - | 5, 6 | 750 | 8860859 |