Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Larp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026235.4 | NP_080511.2 | 2 | 374 | ||
GenBank mRNA | AK017372.1 | - | 2 | 379 | ||
AK020966.1 | - | 2 | 311 | |||
AK039614.1 | - | 2 | 357 | |||
AK042616.1 | - | 2 | 306 | |||
AK087521.1 | - | 2 | 377 | |||
BC090615.1 | - | 2 | 506 | 6809572 |