Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrfap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026242.3 | NP_080518.1 | 1 | 235 | ||
GenBank mRNA | AB041651.1 | - | 1 | 217 | ||
AK005313.1 | - | 1 | 223 | |||
AK009827.1 | - | 1 | 190 | |||
AK017832.1 | - | 1 | 217 | |||
AK018675.1 | - | 1 | 186 | |||
AK020049.1 | - | 1 | 189 | |||
AK159464.1 | - | 1 | 223 | |||
AK167112.1 | - | 1 | 217 | |||
AK170789.1 | - | 1 | 186 | |||
AK170909.1 | - | 1 | 212 | |||
AK170984.1 | - | 1 | 218 | |||
BC010209.1 | - | 1 | 204 | 3482091 | ||
BC085488.1 | - | 1 | 80 | |||
BY227792.1 | - | 1 | 236 |