Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178404.3 | NP_848491.2 | 2 | 484 | ||
GenBank mRNA | AK013610.1 | - | 2 | 480 | ||
AK014766.1 | - | 2 | 478 | |||
AK019345.1 | - | 2 | 480 | |||
AK039171.1 | - | 2 | 479 | |||
AK050503.1 | - | 2 | 485 | |||
BC058173.1 | - | 2 | 449 |