Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042499.1 | NP_001035964.1 | 5 | 536 | ||
GenBank mRNA | AK011280.1 | - | 5 | 478 | ||
AK016099.1 | - | 5 | 545 | |||
AK028195.1 | - | 5 | 530 | |||
AK028269.1 | - | 5 | 523 | |||
AK156982.1 | - | 4 | 4035 | |||
BC050194.1 | - | 5 | 517 | 30022531 |