Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Larp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026235.4 | NP_080511.2 | 3 | 716 | ||
GenBank mRNA | AK017372.1 | - | 3 | 721 | ||
AK020966.1 | - | 3 | 653 | |||
AK035809.1 | - | 293 | ||||
AK039614.1 | - | 3 | 699 | |||
AK042616.1 | - | 3 | 648 | |||
AK047452.1 | - | 1 | 20 | |||
AK087521.1 | - | 3 | 719 | |||
BC090615.1 | - | 3 | 848 | 6809572 |