Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cuta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026307.3 | NP_080583.3 | 5 | 442 | ||
NM_026948.3 | NP_081224.1 | 4 | 390 | |||
GenBank mRNA | AK002828.1 | - | 5 | 386 | ||
AK003381.1 | - | 4 | 330 | |||
AK003647.1 | - | 5 | 384 | |||
AK004197.1 | - | 5 | 384 | |||
AK007580.1 | - | 3 | 519 | |||
AK007912.1 | - | 4 | 465 | |||
AK012608.1 | - | 3 | 758 | |||
AK145185.1 | - | 544 | ||||
AK159336.1 | - | 5 | 389 | |||
AK199328.1 | - | 2 | 49 | |||
AK207031.1 | - | 2 | 49 | |||
AK207668.1 | - | 1 | 124 | |||
BC021463.1 | - | 3 | 504 | |||
BC024422.1 | - | 5 | 372 | 5005082 | ||
BC028531.1 | - | 4 | 481 | |||
BC037500.1 | - | 3 | 368 | |||
CA323205.1 | - | 4 | 356 | |||
CK624897.1 | - | 4 | 367 |