Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026308.2 | NP_080584.1 | 5 | 447 | ||
GenBank mRNA | AJ504717.1 | - | 5 | 454 | ||
AK002780.1 | - | 5 | 448 | |||
AK003455.1 | - | 5 | 445 | |||
AY251016.1 | - | 5 | 469 | |||
AY251017.1 | - | 4 | 368 | |||
BC028491.1 | - | 5 | 457 | 1497187 |