Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpeb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290676.1 | NP_001277605.1 | 9 | 3815 | ||
NM_001290678.1 | NP_001277607.1 | 8 | 3764 | |||
NM_026252.4 | NP_080528.2 | 10 | 3839 | |||
GenBank mRNA | AK015401.1 | - | 6 | 1222 | ||
AK021394.1 | - | 3 | 727 | |||
AK173229.1 | - | 8 | 3773 | |||
BC079599.1 | - | 9 | 1623 |