Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146021.1 | NP_001139493.1 | 3 | 1017 | ||
NM_026253.4 | NP_080529.1 | 3 | 1017 | |||
GenBank mRNA | AK005666.1 | - | 3 | 957 | ||
AK006063.1 | - | 3 | 954 | |||
AK014952.1 | - | 3 | 982 | |||
AK015365.1 | - | 3 | 1021 | |||
AK029909.1 | - | 3 | 982 | |||
AY775400.1 | - | 3 | 1086 | |||
BC048444.1 | - | 3 | 1002 | |||
BC061072.1 | - | 1 | 745 | 6771444 |