Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tctn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026260.2 | NP_080536.2 | 14 | 1649 | ||
GenBank mRNA | BC027335.1 | - | 14 | 1649 | 3710046 | |
BC080805.1 | - | 14 | 1624 | 5352578 | ||
DQ278871.1 | - | 14 | 1666 |