Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpatch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026367.4 | NP_080643.4 | 2 | 330 | ||
GenBank mRNA | AK011727.1 | - | 2 | 315 | ||
AK017975.1 | - | 2 | 299 | |||
AK029990.1 | - | 2 | 315 | |||
AK030026.1 | - | 2 | 329 | |||
AK032734.1 | - | 2 | 305 | |||
AK053781.1 | - | 2 | 322 | |||
AK167826.1 | - | 2 | 334 | |||
AK169660.1 | - | 2 | 331 | |||
BC054810.1 | - | 2 | 302 | 6839419 |