Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026454.3 | NP_080730.1 | 2 | 229 | ||
GenBank mRNA | AK007549.1 | - | 2 | 200 | ||
AK010957.1 | - | 3 | 306 | |||
AK030594.1 | - | 209 | ||||
AK153481.1 | - | 2 | 230 | |||
AK208168.1 | - | 1 | 13 | |||
BC016117.1 | - | 3 | 511 | 4486610 |