Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000202.7 | NP_000193.1 | 3 | 597 | ||
NM_001166550.3 | NP_001160022.1 | 3 | 553 | |||
NM_006123.4 | NP_006114.1 | 3 | 597 | |||
NR_104128.1 | 3 | 597 | ||||
GenBank mRNA | AF050145.1 | - | 3 | 398 | ||
AK294541.1 | - | 3 | 508 | |||
BC006170.1 | - | 3 | 455 | 4025980 | ||
BX647357.1 | - | 3 | 451 | |||
DA325538.1 | - | 3 | 597 | |||
L40586.1 | - | 3 | 507 | |||
M58342.1 | - | 3 | 504 |