Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nop9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026403.3 | NP_080679.3 | 2 | 400 | ||
GenBank mRNA | AK011570.1 | - | 2 | 390 | ||
AK015585.1 | - | 2 | 150 | |||
AK032873.1 | - | 2 | 384 | |||
AK037831.1 | - | 2 | 388 | |||
AK083861.1 | - | 2 | 269 | |||
AK139689.1 | - | 2 | 350 | |||
AK146020.1 | - | 2 | 355 | |||
AK146687.1 | - | 2 | 371 | |||
AK152732.1 | - | 2 | 374 | |||
AK154387.1 | - | 2 | 392 | |||
AK159798.1 | - | 2 | 374 | |||
AK185037.1 | - | 1 | 86 | |||
AK219900.1 | - | 1 | 86 | |||
BC018523.1 | - | 2 | 380 | 3499986 | ||
CJ098414.1 | - | 2 | 401 |