Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026439.2 | NP_080715.2 | 2 | 590 | ||
GenBank mRNA | AB075019.1 | - | 2 | 479 | ||
AK032359.1 | - | 2 | 561 | |||
AK132178.1 | - | 2 | 590 | |||
AK144183.1 | - | 3 | 814 | |||
BC058751.1 | - | 2 | 349 | |||
BC096487.1 | - | 3 | 780 |