Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahctf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026375.2 | NP_080651.2 | 22, 23 | 3483 | ||
GenBank mRNA | AB059278.1 | - | 22, 23 | 3139 | ||
BC138343.1 | - | 22, 23 | 3126 | 8861363 | ||
BC138344.1 | - | 22, 23 | 3126 | 8861364 |