siRNA Details
Gene Symbol: Rnf115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026406.3 | NP_080682.3 | 2 | 336 | ||
GenBank mRNA | AK008329.1 | - | 2 | 190 | ||
AK011584.1 | - | 2 | 266 | |||
AK042216.1 | - | 2 | 276 | |||
AK131966.1 | - | 2 | 256 | |||
AK159772.1 | - | 2 | 229 | |||
BC023113.1 | - | 2 | 283 | 4015975 |

More Information
Other Species Targeted

