Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026434.4 | NP_080710.1 | 3 | 455 | ||
GenBank mRNA | AK008110.1 | - | 3 | 311 | ||
AK021243.1 | - | 3 | 336 | |||
AK035348.1 | - | 3 | 363 | |||
AK036220.1 | - | 2 | 2581 | |||
AK036252.1 | - | 3 | 336 | |||
BC005540.1 | - | 3 | 349 | 3590164 | ||
CB183820.1 | - | 3 | 526 |