Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026454.3 | NP_080730.1 | 6 | 434 | ||
GenBank mRNA | AK007549.1 | - | 6 | 405 | ||
AK007937.1 | - | 6 | 434 | |||
AK007960.1 | - | 6 | 424 | |||
AK010957.1 | - | 7 | 511 | |||
AK030594.1 | - | 414 | ||||
AK153481.1 | - | 6 | 435 | |||
AK208168.1 | - | 5 | 218 | |||
BC016117.1 | - | 7 | 716 | 4486610 |
Gene Symbol: Gm5434 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK136183.1 | - | 1 | 404 | ||
BC085248.1 | - | 1 | 410 |