Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rchy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026557.4 | NP_080833.1 | 1, 2 | 447 | ||
GenBank mRNA | AF071222.1 | - | 1, 2 | 119 | ||
AF276959.1 | - | 1, 2 | 117 | |||
AF305423.1 | - | 1, 2 | 103 | |||
AK018364.1 | - | 1, 2 | 114 | |||
AK018488.1 | - | 1, 2 | 144 | |||
AK078397.1 | - | 1, 2 | 231 | |||
BC023138.1 | - | 1, 2 | 221 | 4008685 | ||
BC057143.1 | - | 1, 2 | 447 | 5249543 |