Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdccag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085407.1 | NP_001078876.1 | 2, 3 | 245 | ||
NM_026563.3 | NP_080839.2 | 3, 4 | 395 | |||
GenBank mRNA | AK014885.1 | - | 3, 4 | 364 | ||
AK021273.1 | - | 3, 4 | 386 | |||
AK082787.1 | - | 2, 3 | 216 | |||
BC061037.1 | - | 2, 3 | 212 |