Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tppp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026481.3 | NP_080757.1 | 4 | 745 | ||
GenBank mRNA | AK003376.1 | - | 1 | 27 | ||
AK009771.1 | - | 4 | 709 | |||
AK012437.1 | - | 4 | 713 | |||
AK155018.1 | - | 4 | 716 | |||
AK160507.1 | - | 4 | 716 | |||
BC010788.1 | - | 4 | 702 | 4216285 | ||
BC116622.1 | - | 3 | 537 |