Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tctn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026486.3 | NP_080762.1 | 11 | 1421 | ||
GenBank mRNA | AK012720.1 | - | 11 | 1395 | ||
AK014476.1 | - | 11 | 1387 | |||
AK031829.1 | - | 11 | 1422 | |||
AK032378.1 | - | 10 | 1506 | |||
AK087856.1 | - | 11 | 1380 | |||
AK162182.1 | - | 1179 | ||||
BC058375.1 | - | 11 | 1284 | 6828178 | ||
DQ278869.1 | - | 11 | 1419 |